
4'-Chloroacetophenone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-91-2
:
Formula:C₈H₇ClO
MDL Number:MFCD00000624
:154.6 g/mol
Merck Index:13,02133
Boiling Pt:232 °C (1013 hPa)
Melting Pt:18…20 °C
Density:1.1952 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-800-7
UN:3416
ADR:6.1,II
:M-Clarity™ quality level = MQ100
Flash Pt:>100 °C
:GHS07
Frequently Bought Together