Order Entry
Czech Republic
ContactUsLinkComponent
4'-Chloroacetophenone 98+%
Irritant
4'-Chloroacetophenone 98+%
Certificates
undefined
Irritant
4'-Chloroacetophenone 98+%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5539 to 1.5559 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Specific optical rotation:
    -7° to -8.5° (20°C, 589 nm) (c=1.13, MeOH)

Specification Test Results

Related Information

Frequently Bought Together