
3'-Chloroacetophenone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-02-5
Formula:C₈H₇ClO
MDL Number:MFCD00000593
:154.6 g/mol
Boiling Pt:241 °C (1013 hPa)
Density:1.184 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-721-8
UN:3416
ADR:6.1,II
:M-Clarity™ quality level = MQ100
:GHS07
Frequently Bought Together