Order Entry
Czech Republic
ContactUsLinkComponent
 
ω,2,4-Trichloroacetophenone
Catalog # EHERC17690000
Supplier:  EHRENSTORFER
CAS Number:  
ω,2,4-Trichloroacetophenone
Catalog # EHERC17690000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :223.49 g/mol
Boiling Pt:292 °C (1013 hPa)
Melting Pt:58…60 °C
Storage Temperature:Ambient
EINECS:224-218-2
UN:2923
ADR:8,III
CAS Number:4252-78-2
Formula:C₈H₅Cl₃O
MDL Number:MFCD00000934