Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
ω,2,4-Trichloroacetophenone ≥97%

ω,2,4-Trichloroacetophenone ≥97%

Toxic
Corrosive
Irritant
ω,2,4-Trichloroacetophenone ≥97%
Certificates
ω,2,4-Trichloroacetophenone ≥97%
ω,2,4-Trichloroacetophenone ≥97%

3 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 4252-78-2 Molecular Formula: C8H5Cl3O Formula Weight: 223.49 Flash Point: 110°(230°F) Melting Point: 52-56° Beilstein Reference: 957098 UN No.: 2923 MDL No.: MFCD00000934

Synonyms:  2,4-Dichlorophenacyl chloride, α,2,4-Trichloroacetophenone, 2,2',4'-Trichloroacetophenone
  • CAS Number:4252-78-2
  • Formula:C₈H₅Cl₃O
  • MDL Number:MFCD00000934
  • Molecular Weight:223.49 g/mol
  • Boiling Pt:292 °C (1013 hPa)
  • Melting Pt:52…56 °C
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:224-218-2
  • UN:2923
  • ADR:8,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.