

(4-Chlorophenyl)acetonitrile 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:140-53-4
:
Formula:C₈H₆ClN
MDL Number:MFCD00001918
:151.6 g/mol
Boiling Pt:141…143 °C (11 mmHg)
Melting Pt:28…31 °C
Density:1.190
Flash Pt:134 °C (273 °F)
Storage Temperature:Ambient
EINECS:205-418-9
UN:3439
ADR:6.1,II
:GHS06
:GHS07
Related Information
Frequently Bought Together