

(2-Chlorophenyl)acetonitrile ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2856-63-5
:
Formula:C₈H₆ClN
MDL Number:MFCD00001898
Boiling Pt:240…242 °C
Melting Pt:21…24 °C
Density:1 g/cm³ (20 °C)
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:220-669-4
ADR:6.1,III
:151.6 g/mol
UN:3439
:GHS06
:GHS07
Related Information
Frequently Bought Together