

2-chlortoluen, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:95-49-8
:
Formula:C₇H₇Cl
MDL Number:MFCD00000562
Merck Index:13,02190
Boiling Pt:159 °C (1013 hPa)
Melting Pt:–36 °C
Density:1,0825 g/cm³ (20 °C)
Flash Pt:48 °C
UN:2238
ADR:3,III
:M-Clarity™ quality level = MQ100
EINECS:202-424-3
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together