
4-Bromo-m-cresol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14472-14-1
:
Formula:C₇H₇BrO
MDL Number:MFCD00079723
:187.04 g/mol
Boiling Pt:142…145 °C (23 mmHg)
Melting Pt:57…61 °C
Storage Temperature:Ambient
EINECS:238-464-3
:GHS07
Related Information
Frequently Bought Together