
2-Bromo-p-cresol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6627-55-0
:
Formula:C₇H₇BrO
MDL Number:MFCD00002151
:187.04 g/mol
Boiling Pt:213…214 °C
Melting Pt:16 °C
Density:1.547
Flash Pt:>110 °C (230 °F)
EINECS:229-595-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together