
2-Bromo-6-nitrotoluene, Sigma-Aldrich®
Catalog # SIAL260053-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:216.03 g/mol
Boiling Pt:143 °C (22 mmHg)
Melting Pt:42 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:259-566-4
:GHS07
CAS Number:55289-35-5
Formula:C₇H₆BrNO₂
MDL Number:MFCD00009792