ω-Bromo-4-fluoroacetophenone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:403-29-2
:
Formula:C₈H₆BrFO
MDL Number:MFCD00040830
:217.04 g/mol
Boiling Pt:82 °C (0.4 mmHg)
Melting Pt:49 °C
:M-Clarity™ quality level = MQ100
EINECS:206-955-1
Frequently Bought Together