Order Entry
Czech Republic
ContactUsLinkComponent
α-Bromo-3,4-difluorotoluene, Sigma-Aldrich®
α-Bromo-3,4-difluorotoluene, Sigma-Aldrich®
Catalog # SIAL264458-5G
Supplier:  Merck
CAS Number:  
α-Bromo-3,4-difluorotoluene, Sigma-Aldrich®
Catalog # SIAL264458-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :207.02 g/mol
Boiling Pt:59…61 °C (3.5 mmHg)
Density:1.61 g/cm³ (20 °C)
ADR:8,II
 :M-Clarity™ quality level = MQ200
EINECS:285-653-1
UN:3265
CAS Number:85118-01-0
Formula:C₇H₅BrF₂
MDL Number:MFCD00009889