ω-Bromo-2,5-dimethoxyacetophenone, Sigma-Aldrich®
Catalog # SIAL104582-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:259.1 g/mol
Boiling Pt:315 °C (1013 hPa)
Melting Pt:91 °C
:M-Clarity™ quality level = MQ100
EINECS:214-873-2
CAS Number:1204-21-3
Formula:C₁₀H₁₁BrO₃
MDL Number:MFCD00000198