Order Entry
Czech Republic
ContactUsLinkComponent
ω-Bromo-2,4-dimethoxyacetophenone 98%
Corrosive
ω-Bromo-2,4-dimethoxyacetophenone 98%
Catalog # ACRO190010010
CAS Number:  
undefined
Corrosive
ω-Bromo-2,4-dimethoxyacetophenone 98%
Catalog # ACRO190010010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    101 to 105 °C
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :259.1 g/mol
Boiling Pt:315 °C (1013 hPa)
Melting Pt:101…105 °C
EINECS:262-542-6
UN:3261
ADR:8,III
 :GHS05
CAS Number:60965-26-6
Formula:C₁₀H₁₁BrO₃
MDL Number:MFCD00000197