
4'-Bromoacetophenone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-90-1
:
Formula:C₈H₇BrO
MDL Number:MFCD00000105
:199.05 g/mol
Merck Index:13,01387
Boiling Pt:255 °C (1013 hPa)
Melting Pt:49…52 °C
Density:1.736 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-799-3
UN:2811
ADR:6.1,II
:M-Clarity™ quality level = MQ200
Flash Pt:>100 °C
:GHS07
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:49 - 52 °C
- Molar Mass:199.04 g/mol
- Boiling Point:255 °C (1013 hPa)
- Flash Point:100 °C
Specification Test Results
Frequently Bought Together