ω,3-Dibromoacetophenone, Sigma-Aldrich®
Catalog # SIAL654787-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Melting Pt:51 °C
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ100
:277.94 g/mol
CAS Number:18523-22-3
Formula:C₈H₆Br₂O
MDL Number:MFCD00082698