
1,2-Dibromo-1-phenylethane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:93-52-7
:
Formula:C₆H₅CHBrCH₂Br
MDL Number:MFCD00000138
:263.96 g/mol
Boiling Pt:139…141 °C (15 mmHg)
Melting Pt:75…76 °C
Density:1.74 g/cm³ (20 °C)
EINECS:202-253-4
ADR:8,III
:M-Clarity™ quality level = MQ100
UN:3261
:GHS05
Storage Temperature:Ambient
Frequently Bought Together