cis-Stilbene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:645-49-8
:
Formula:(C₆H₅)CHCH(C₆H₅)
MDL Number:MFCD00004788
:180.25 g/mol
Merck Index:12,08972
Boiling Pt:307 °C (1013 hPa)
Density:1.01 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:211-445-7
Frequently Bought Together