4-Allyl-2,6-dimethoxyphenol, Sigma-Aldrich®
Catalog # SIALA31601-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:194.23 g/mol
:M-Clarity™ quality level = MQ100
EINECS:229-600-2
CAS Number:6627-88-9
Formula:C₁₁H₁₄O₃
MDL Number:MFCD00008655