Order Entry
Czech Republic
ContactUsLinkComponent
Phloroglucinol for synthesis, Sigma-Aldrich®
Irritant
Phloroglucinol for synthesis, Sigma-Aldrich®
Catalog # 8.18887.0100
Supplier:  Merck
CAS Number:  
Irritant
Phloroglucinol for synthesis, Sigma-Aldrich®
Catalog # 8.18887.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    10 g/l (20 °C)
  • Melting Point:
    217 - 220 °C
  • Molar Mass:
    126.11 g/mol
  • Bulk Density:
    560 kg/m3

Specification Test Results

About this item

 :
 :126.11 g/mol
Merck Index:12,07482
Boiling Pt:400 °C (1013 hPa)
Melting Pt:217…220 °C
Density:1.46 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:203-611-2
 :GHS07
CAS Number:108-73-6
Formula:C₆H₆O₃
MDL Number:MFCD00002286