Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Floroglucinol
Floroglucinol, anhydrous 98%
Floroglucinol, anhydrous 98%
:
Thermo Fisher Scientific
Floroglucinol, anhydrous 98%
:
Thermo Fisher Scientific
:
108-73-6
:
:
C₆H₆O₃
:
MFCD00002286
:
14,7328
:
400 °C (1013 hPa)
:
215…220 °C
:
1,46 g/cm³ (20 °C)
:
Ambient
:
203-611-2
:
GHS07
ALFAB25502.18_TDS_EN
msds_ALFAB25502
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Aceton, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
: Merck
Aceton ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Dichlormethan stabilised, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
: VWR Chemicals
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Xylen (směs izomerů) ≥98%, TECHNICAL
: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitril, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® USP, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
L(+)-cystein
: Apollo Scientific