
Phloroglucinol, anhydrous 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-73-6
:
Formula:C₆H₆O₃
MDL Number:MFCD00002286
:126.11 g/mol
Merck Index:12,07482
Boiling Pt:400 °C (1013 hPa)
Melting Pt:215…218 °C
Density:1,46 g/cm³ (20 °C)
EINECS:203-611-2
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:213 to 218 °C
- Loss on drying:≤2 % (1 g, 120 °C)
- HPLC:≥99.0 %
Specification Test Results
Frequently Bought Together