

Pyrogallol ≥99.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 27206.231
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay:Min. 99.5 %
- Acidity:Max. 0.005 meq/g
- Melting point:131 - 134 °C
- Heavy metals (as Pb):Max. 5 ppm
- Ignition residue (SO4):Max. 50 ppm
- Cl (Chloride):Max. 10 ppm
- SO4 (Sulphate):Max. 50 ppm
- Fe (Iron):Max. 10 ppm
Specification Test Results
About this item
:
:126.11 g/mol
Merck Index:13,08090
Boiling Pt:309 °C (1013 hPa)
Melting Pt:130…132 °C
Density:1.453 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-762-9
UN:2811
ADR:6.1,III
:GHS08
:GHS07
CAS Number:87-66-1
Formula:C₆H₆O₃
MDL Number:MFCD00002192