Order Entry
Czech Republic
ContactUsLinkComponent
Pyrogallol for synthesis, Sigma-Aldrich®
Pyrogallol for synthesis, Sigma-Aldrich®
Catalog # 8.22302.0250
Supplier:  Merck
CAS Number:  
undefined
Pyrogallol for synthesis, Sigma-Aldrich®
Catalog # 8.22302.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    625 g/l (25 °C)
  • Melting Point:
    131 - 135 °C
  • Molar Mass:
    126.11 g/mol
  • Bulk Density:
    600 kg/m3
  • Boiling Point:
    309 °C (1013 hPa)
  • Vapor Pressure:
    2 - 4 hPa (140 °C)
  • Density:
    1.45 g/cm3 (20 °C)
  • pH:
    4 - 5 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

Specification Test Results

About this item

 :
 :126.11 g/mol
Merck Index:13,08090
Boiling Pt:309 °C (1013 hPa)
Melting Pt:130…132 °C
Density:1.453 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-762-9
UN:2811
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
 :GHS07
 :GHS08
CAS Number:87-66-1
Formula:C₆H₆O₃
MDL Number:MFCD00002192