Hydroquinone 99.5%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 1,4-Dihydroxybenzene, Quinol
- CAS Number:123-31-9
- Formula:C₆H₄(OH)₂
- MDL Number:MFCD00002339
- Molecular Weight:110.11 g/mol
- Merck Index:13,04833
- Boiling Pt:285…287 °C (760 mmHg)
- Melting Pt:170…174 °C
- Density:1,332 g/cm³ (20 °C)
- Flash Pt:165 °C
- Storage Temperature:Ambient
- EINECS:204-617-8
- UN:3077
- ADR:9,III
5 Options Available Below
Product Details & Documents
Synonyms: 1,4-Dihydroxybenzene, Quinol
- CAS Number:123-31-9
- Formula:C₆H₄(OH)₂
- MDL Number:MFCD00002339
- Molecular Weight:110.11 g/mol
- Merck Index:13,04833
- Boiling Pt:285…287 °C (760 mmHg)
- Melting Pt:170…174 °C
- Density:1,332 g/cm³ (20 °C)
- Flash Pt:165 °C
- Storage Temperature:Ambient
- EINECS:204-617-8
- UN:3077
- ADR:9,III
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:171 to 174 °C
- Water:≤0.6 % (K.F.)
- Sulfated ash:≤0.05 %
- HPLC:≥99.4 %
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



