Order Entry
Czech Republic
ContactUsLinkComponent
 
Pyrocatechol 99+%
 
undefined
Pyrocatechol 99+%
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Off-white to grey-brown Crystalline flakes (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Melting point:
    103 to 106 °C
  • GC:
    ≥99.0 %
  • Heavy metals (as Pb):
    ≤10 ppm
  • Insoluble matter:
    ≤0.05 % (in water)

 

 

Frequently Bought Together