1,3-Dimethoxybenzene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:151-10-0
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00008384
:138.17 g/mol
Boiling Pt:218 °C (1013 hPa)
Melting Pt:–52 °C
Density:1.05 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:205-783-4
Frequently Bought Together