Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
2-Phenoxyethanol

2-Phenoxyethanol

Glass bottle for solids, 500 mg
Catalog # PROOMM0952.00
Irritant
2-Phenoxyethanol
Catalog # PROOMM0952.00
CAS Number:  122-99-6
2-Phenoxyethanol
2-Phenoxyethanol

Glass bottle for solids, 500 mg

CAS Number:  122-99-6
Price
40 630,00 Kč
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
  • CAS Number:122-99-6
  • Formula:C₆H₅OCH₂CH₂OH
  • MDL Number:MFCD00002857
  • Molecular Weight:138.17 g/mol
  • Merck Index:12,07410
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:13 °C
  • Density:1.1094 g/cm³ (20 °C)
  • Flash Pt:121 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:204-589-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg

Specifications