Order Entry
Czech Republic
ContactUsLinkComponent
2-Phenoxyethanol 99.0-100.5% Ph. Eur.
Irritant
2-Phenoxyethanol 99.0-100.5% Ph. Eur.
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
2-Phenoxyethanol 99.0-100.5% Ph. Eur.
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    99.0 - 100.5 %
  • Appearance:
    C'less slightly visc.liq.
  • Identification (IR):
    Passes test
  • Relative density:
    1.105 - 1.11
  • Unspecified impurities (for each):
    Max. 0.10 %
  • Total impurities:
    Max. 0.3 %
  • Reporting treshold:
    Max. 0.05 %
  • Residual solvents:
    Passes test
  • As (Arsenic) :
    ≤ 1 ppm
  • Cd (Cadmium) :
    ≤ 1 ppm
  • Co (Cobalt) :
    ≤ 1 ppm
  • Hg (Mercury) :
    ≤ 1 ppm
  • Ni (Nickel) :
    ≤ 1 ppm
  • Pb (Lead) :
    ≤ 1 ppm
  • V (Vanadium) :
    ≤ 1 ppm
  • Conforms to Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together