
2-Phenoxyethanol for synthesis, Sigma-Aldrich®
: Merck
:122-99-6
:
:C₆H₅OCH₂CH₂OH
:MFCD00002857
:138.17 g/mol
:12,07410
:245 °C (1013 hPa)
:13 °C
:1.1094 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:121 °C (closed cup)
:204-589-7
:GHS07
:Ambient
- Solubility:24 g/l (20 °C)
- Melting Point:13 °C
- Molar Mass:138.16 g/mol
- Boiling Point:244 - 250 °C (1013 hPa)
- Vapor Pressure:0.04 hPa (20 °C)
- Flash Point:121 °C DIN 51758
- Refractive Index:1.534 (20 °C, 589 nm)
- Explosion Limit:1.4 - 9.0 %(V)
- Density:1.11 g/cm3 (20 °C)
- pH:7 (10 g/l, H2O, 23 °C)
- Ignition Point:535 °C DIN 51794
Frequently Bought Together