
2-Phenoxyethanol
Catalog # EHERC13328300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:138.17 g/mol
Merck Index:12,07410
Boiling Pt:245 °C (1013 hPa)
Melting Pt:13 °C
Density:1.1094 g/cm³ (20 °C)
Flash Pt:121 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-589-7
:GHS07
CAS Number:122-99-6
Formula:C₆H₅OCH₂CH₂OH
MDL Number:MFCD00002857