
2-Phenoxyethanol ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:122-99-6
:
Formula:C₆H₅OCH₂CH₂OH
MDL Number:MFCD00002857
:138.17 g/mol
Merck Index:14,7257
Boiling Pt:245 °C
Melting Pt:11…14 °C
Density:1.102
Flash Pt:121 °C (249 °F)
EINECS:204-589-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together