
2-Nitrobenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1694-92-4
:
Formula:C₆H₄ClNO₄S
MDL Number:MFCD00007430
Melting Pt:69.5…69.7 °C
Storage Temperature:Ambient
ADR:8,II
:M-Clarity™ quality level = MQ100
:221.62 g/mol
EINECS:216-907-1
UN:3261
:GHS05
Frequently Bought Together