
2-Nitrobenzenesulphonyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1694-92-4
:
Formula:C₆H₄ClNO₄S
MDL Number:MFCD00007430
Melting Pt:64…69 °C
Storage Temperature:Ambient
EINECS:216-907-1
UN:3261
ADR:8,II
:221.62 g/mol
:GHS05
Related Information
Frequently Bought Together