
Barium diphenylamine-4-sulphonate, pure indicator grade
:6211-24-1
:1300-92-1
:C₂₄H₂₀BaN₂O₆S₂
:MFCD00007497
:633.89 g/mol
:300 °C
:Ambient
:228-278-0
:1564
:6.1,III
:GHS07
- Appearance:Off-white to grey Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 % (105°C)
- Sensitivity:(as indicator) Passes test
Frequently Bought Together