Difenylamin 99%, pure
:122-39-4
:
:(C₆H₅)₂NH
:MFCD00003014
:13,03349
:302 °C (760 mmHg)
:52…54 °C
:1,165 g/cm³ (20 °C)
:152 °C
:Ambient
:204-539-4
:3077
:9,III
:GHS06
:GHS08
:GHS09
- Appearance:White to yellow to beige Flakes
- Infrared spectrum:Conforms
- Melting point:52 to 55 °C
- GC:≥98.5 %
- Free acid:≤1 %
Frequently Bought Together