
Sulphanilic acid ≥98%, GPR RECTAPUR®
Catalog # 20673.294
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:Min. 98 %
- Ignition residue (SO4):Max. 0.1 %
- Cl (Chloride):Max. 100 ppm
- SO4 (Sulphate):Max. 0.2 %
Specification Test Results
About this item
:
:173.19 g/mol
Merck Index:13,09012
Boiling Pt:363 °C (1013 hPa)
Melting Pt:279 °C
Density:1.485 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:204-482-5
:GHS07
CAS Number:121-57-3
Formula:4-(H₂N)C₆H₄SO₃H
MDL Number:MFCD00007886