

4-Methoxyphenyl isocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5416-93-3
:
Formula:C₈H₇NO₂
MDL Number:MFCD00002026
:149.15 g/mol
Boiling Pt:119…120 °C (20 mmHg)
Density:1.151
Flash Pt:98 °C (208 °F)
EINECS:226-513-1
UN:2206
ADR:6.1,III
:GHS08
:GHS07
Related Information
Frequently Bought Together