
4-Methoxyphenyl isocyanate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5416-93-3
:
Formula:C₈H₇NO₂
MDL Number:MFCD00002026
:149.15 g/mol
Boiling Pt:106…110 °C (16 mmHg)
Density:1.15 g/cm³
Flash Pt:98 °C
EINECS:226-513-1
UN:2206
ADR:6.1,II
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5450 to 1.5470 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together