
N,N,N',N'-Tetramethyl-p-phenylenediamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-22-1
:
Formula:C₁₀H₁₆N₂
MDL Number:MFCD00008309
:164.25 g/mol
Merck Index:12,09367
Boiling Pt:260 °C (1013 hPa)
Melting Pt:48…50 °C
Density:1.08 g/cm³ (20 °C)
UN:2811
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:202-831-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:48 - 50 °C
- Molar Mass:164.24 g/mol
- Boiling Point:260 °C
- Flash Point:112 °C
Specification Test Results
Frequently Bought Together