3-(Dimethylamino)phenol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-07-0
:
Formula:C₈H₁₁NO
MDL Number:MFCD00002264
:137.18 g/mol
Boiling Pt:265 °C (1013 hPa)
Melting Pt:65…72 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:202-727-0
Frequently Bought Together