Azobenzene for synthesis, Sigma-Aldrich®
: Merck
:103-33-3
:
:C₁₂H₁₀N₂
:MFCD00003022
:182.22 g/mol
:12,00948
:293 °C (1013 hPa)
:66…69 °C
:1.203 g/cm³ (20 °C)
:203-102-5
:3077
:9,III
:M-Clarity™ quality level = MQ200
:140 °C
:GHS08
:GHS07
:GHS09
:Ambient
- Solubility:6.4 mg/l (25 °C)
- Melting Point:66 - 69 °C
- Molar Mass:182.23 g/mol
- Boiling Point:291 - 294 °C (1013 hPa)
- Vapor Pressure:0.00048 hPa (25 °C)
- Flash Point:100 °C
- Density:1.203 g/cm3
Frequently Bought Together