


Azobenzene
Catalog # EHERC10390000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
:182.22 g/mol
Merck Index:12,00948
Boiling Pt:293 °C (1013 hPa)
Melting Pt:66…69 °C
Density:1.203 g/cm³ (20 °C)
Flash Pt:140 °C
EINECS:203-102-5
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:103-33-3
Formula:C₁₂H₁₀N₂
MDL Number:MFCD00003022