
1-Iodo-2,4-dinitrobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:709-49-9
:
Formula:C₆H₃IN₂O₄
MDL Number:MFCD00039738
:294.01 g/mol
Melting Pt:87…90 °C
EINECS:211-908-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together