Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-4-iodobenzoate 98%
Environment
Methyl-4-iodobenzoate 98%
Certificates
undefined
Environment
Methyl-4-iodobenzoate 98%

Specification Test Results

  • Appearance:
    White to beige to light orange Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    113 to 117 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together