Order Entry
Czech Republic
ContactUsLinkComponent
4-Fluorobenzenesulphonyl chloride 98%
Corrosive
4-Fluorobenzenesulphonyl chloride 98%
Certificates
undefined
Corrosive
4-Fluorobenzenesulphonyl chloride 98%

Specification Test Results

  • Appearance:
    White to light brown Low melting crystalline mass
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥97.5 % (ex Cl)
  • GC:
    ≥98.5 %
  • Iron (Fe):
    ≤0.0005 %

Specification Test Results

Related Information

Frequently Bought Together