
2,6-Difluorobenzenesulphonyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60230-36-6
:
Formula:C₆H₃ClF₂O₂S
MDL Number:MFCD00085004
:212.6 g/mol
Boiling Pt:127…130 °C (6.5 mmHg)
Density:1.568
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Related Information
Frequently Bought Together