
2,4-Difluorobenzenesulphonyl chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13918-92-8
:
Formula:C₆H₃ClF₂O₂S
MDL Number:MFCD00042481
:212.6 g/mol
Boiling Pt:101…102 °C (1 mmHg)
Density:1.581 g/cm³
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5165 to 1.5185 (20°C, 589 nm)
- Titration Argentometric:≥97.5 % (ex Cl)
Specification Test Results
Frequently Bought Together