1,3-Dimethoxy-5-fluorobenzene, Sigma-Aldrich®
Catalog # SIAL322342-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:156.16 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:52189-63-6
Formula:C₈H₉FO₂
MDL Number:MFCD00012445